Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF18
Entrez Gene ID
7566
Gene Name
zinc finger protein 18
Gene Aliases
HDSG1, KOX11, ZKSCAN6, ZNF535, ZSCAN38, Zfp535
Chromosome Location
Chr.17: 11957433 - 11997510 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303281.1 NP_001290210.1 6 - 7 1019 111
NM_001303282.1 NP_001290211.1 6 - 7 1016 111
NM_144680.3 NP_653281.2 8 - 9 1462 111
XM_011524002.2 XP_011522304.1 6 - 7 1092 111
XM_017025006.1 XP_016880495.1 6 - 7 1019 111
XM_017025007.1 XP_016880496.1 7 - 8 1362 111
XM_017025008.1 XP_016880497.1 6 - 7 1016 111
XM_017025009.1 XP_016880498.1 5 - 6 1344 111
XM_017025010.1 XP_016880499.1 6 - 7 1089 111
XM_017025011.1 XP_016880500.1 7 - 8 1359 111
GenBank mRNA AK092656.1 - 8 - 9 1462 111
AK127908.1 - 6 - 7 977 111
AY491053.1 - 6 - 7 963 111
BC036096.2 - 5 - 6 940 111

View Full Product Details