The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320663.1 | NP_001307592.1 | 4 - 5 | 1641 | 74 |
NM_001649.3 | NP_001640.1 | 8 - 9 | 4406 | 74 | |
XM_011545506.1 | XP_011543808.1 | 4 - 5 | 1518 | 74 | |
XM_017029493.1 | XP_016884982.1 | 4 - 5 | 2011 | 74 | |
GenBank mRNA | AK096596.1 | - | 4 - 5 | 1972 | 74 |
AK299907.1 | - | 4 - 5 | 1641 | 74 | |
BC140866.1 | - | 8 - 9 | 4367 | 74 | |
X83543.1 | - | 8 - 9 | 4406 | 74 |