The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006198.2 | NP_006189.2 | 1 - 2 | 93 | 81 |
GenBank mRNA | AF416716.1 | - | 1 - 2 | 41 | 81 |
AK289964.1 | - | 1 - 2 | 76 | 81 | |
BC013791.1 | - | 1 - 2 | 69 | 81 | |
U52969.1 | - | 1 - 2 | 93 | 81 | |
X93349.1 | - | 1 - 2 | 70 | 81 |