The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006198.2 | NP_006189.2 | 2 - 3 | 142 | 86 |
GenBank mRNA | AF416716.1 | - | 2 - 3 | 90 | 86 |
AK289964.1 | - | 2 - 3 | 125 | 86 | |
AM183336.1 | - | 2 - 3 | 80 | 86 | |
BC013791.1 | - | 2 - 3 | 118 | 86 | |
CR450319.1 | - | 1 - 2 | 61 | 86 | |
U52969.1 | - | 2 - 3 | 142 | 86 |