The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316350.1 | NP_001303279.1 | 7 - 8 | 1073 | 79 |
NM_014236.3 | NP_055051.1 | 8 - 9 | 1256 | 79 | |
XM_005273313.4 | XP_005273370.1 | 8 - 9 | 1216 | 79 | |
XM_011544303.2 | XP_011542605.1 | 9 - 10 | 1305 | 79 | |
XM_011544304.2 | XP_011542606.1 | 8 - 9 | 1124 | 79 | |
GenBank mRNA | AB209882.1 | - | 8 - 9 | 1161 | 79 |
AF035285.1 | - | 2 - 3 | 159 | 79 | |
AF043937.1 | - | 8 - 9 | 1132 | 79 | |
AJ002190.1 | - | 8 - 9 | 1207 | 79 | |
AK222773.1 | - | 8 - 9 | 1212 | 79 | |
AK223304.1 | - | 8 - 9 | 1215 | 79 | |
AK297982.1 | - | 7 - 8 | 1034 | 79 | |
AK311386.1 | - | 7 - 8 | 906 | 79 | |
AK314181.1 | - | 8 - 9 | 1219 | 79 | |
BC000450.2 | - | 8 - 9 | 1103 | 79 |