The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145998.1 | NP_001139470.1 | 4 - 5 | 590 | 87 |
NM_021082.3 | NP_066568.3 | 5 - 6 | 683 | 87 | |
XM_005247722.3 | XP_005247779.1 | 5 - 6 | 567 | 87 | |
XM_006713736.3 | XP_006713799.1 | 5 - 6 | 567 | 87 | |
XM_017007074.1 | XP_016862563.1 | 5 - 6 | 567 | 87 | |
GenBank mRNA | AK289820.1 | - | 5 - 6 | 567 | 87 |
AK302002.1 | - | 4 - 5 | 477 | 87 | |
AK304189.1 | - | 4 - 5 | 475 | 87 | |
BC020743.1 | - | 5 - 6 | 567 | 87 | |
BC038445.1 | - | 5 - 6 | 566 | 87 | |
BC044572.1 | - | 5 - 6 | 567 | 87 | |
S78203.1 | - | 5 - 6 | 560 | 87 |