The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003014.3 | NP_003005.2 | 4 - 5 | 126 | |
GenBank mRNA | AF026692.1 | - | 4 - 5 | 1055 | 126 |
AK302357.1 | - | 4 - 5 | 126 | ||
AK303977.1 | - | 5 - 6 | 1004 | 126 | |
AK313029.1 | - | 4 - 5 | 1074 | 126 | |
BC047684.1 | - | 4 - 5 | 126 | ||
BC058911.1 | - | 4 - 5 | 1062 | 126 | |
BT019679.1 | - | 4 - 5 | 126 | ||
CR541731.1 | - | 4 - 5 | 798 | 126 | |
CR541755.1 | - | 4 - 5 | 798 | 126 | |
DQ420628.1 | - | 4 - 5 | 126 |