The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308419.1 | NP_001295348.1 | 2 - 3 | 204 | 60 |
NM_022165.2 | NP_071448.1 | 2 - 3 | 204 | 60 | |
GenBank mRNA | AF311862.1 | - | 2 - 3 | 182 | 60 |
AY358744.1 | - | 2 - 3 | 204 | 60 | |
BC027618.2 | - | 2 - 3 | 178 | 60 | |
BM069859.1 | - | 2 - 3 | 161 | 60 |