The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135098.1 | NP_001128570.1 | 6 - 7 | 635 | 131 |
NM_173355.3 | NP_775491.1 | 4 - 5 | 638 | 131 | |
XM_005246359.3 | XP_005246416.1 | 4 - 5 | 638 | 131 | |
XM_017003484.1 | XP_016858973.1 | 4 - 5 | 638 | 131 | |
GenBank mRNA | AK122743.2 | - | 4 - 5 | 638 | 131 |
BC033529.2 | - | 6 - 7 | 635 | 131 | |
KU178873.1 | - | 4 - 5 | 444 | 131 | |
KU178874.1 | - | 4 - 5 | 444 | 131 | |
KU178875.1 | - | 3 - 4 | 285 | 131 |