The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032408.3 | NP_115784.1 | 11 - 12 | 3421 | 89 |
XM_017012773.1 | XP_016868262.1 | 11 - 12 | 3434 | 89 | |
GenBank mRNA | AB032253.1 | - | 11 - 12 | 3414 | 89 |
AF084479.1 | - | 11 - 12 | 3353 | 89 | |
BC136520.1 | - | 11 - 12 | 3254 | 89 |