Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BAZ1B
Entrez Gene ID
9031
Gene Name
bromodomain adjacent to zinc finger domain 1B
Gene Aliases
WBSCR10, WBSCR9, WSTF
Chromosome Location
Chr.7: 73440398 - 73522298 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032408.3 NP_115784.1 17 - 18 4344 60
XM_017012773.1 XP_016868262.1 17 - 18 4357 60
GenBank mRNA AB032253.1 - 17 - 18 4337 60
AF072810.1 - 17 - 18 4344 60
AF084479.1 - 17 - 18 4276 60
BC136520.1 - 17 - 18 4177 60

View Full Product Details