Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KYNU
Entrez Gene ID
8942
Gene Name
kynureninase
Gene Aliases
KYNUU
Chromosome Location
Chr.2: 142877626 - 143042316 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032998.1 NP_001028170.1 4 - 5 503 104
NM_001199241.1 NP_001186170.1 5 - 6 589 104
NM_003937.2 NP_003928.1 4 - 5 503 104
XM_017005217.1 XP_016860706.1 4 - 5 465 104
GenBank mRNA AK223426.1 - 4 - 5 501 104
AK290900.1 - 4 - 5 449 104
AK315343.1 - 5 - 6 551 104
AL548658.3 - 4 - 5 631 104
BC000879.1 - 4 - 5 472 104
CB151988.1 - 4 - 5 503 104
CR457423.1 - 3 - 4 373 104
U57721.1 - 4 - 5 479 104

View Full Product Details