The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278270.1 | NP_001265199.1 | 7 - 8 | 1143 | 82 |
NM_006354.3 | NP_006345.1 | 7 - 8 | 1749 | 82 | |
NM_133480.2 | NP_597814.1 | 7 - 8 | 1749 | 82 | |
NR_103488.1 | 5 - 6 | 1185 | 82 | ||
GenBank mRNA | AF069733.1 | - | 6 - 7 | 926 | 82 |
AK000228.1 | - | 7 - 8 | 1153 | 82 | |
AK024982.1 | - | 3 - 4 | 1105 | 82 | |
AK292264.1 | - | 7 - 8 | 1143 | 82 | |
AL117487.1 | - | 7 - 8 | 1412 | 82 | |
BC009240.2 | - | 7 - 8 | 1474 | 82 | |
BC013433.1 | - | 7 - 8 | 1412 | 82 | |
BG477633.1 | - | 6 - 7 | 447 | 82 | |
CR533543.1 | - | 6 - 7 | 926 | 82 |