The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004759.4 | NP_004750.1 | 9 - 10 | 1265 | 73 |
XM_005273353.3 | XP_005273410.1 | 9 - 10 | 1373 | 73 |