Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLMN
Entrez Gene ID
11146
Gene Name
glomulin, FKBP associated protein
Gene Aliases
FAP, FAP48, FAP68, FKBPAP, GLML, GVM, VMGLOM
Chromosome Location
Chr.1: 92245786 - 92441936 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053274.2 NP_444504.1 11 - 12 1217 145
NR_135089.1 11 - 12 1217 145
XM_005270401.3 XP_005270458.1 10 - 11 1079 145
XM_006710309.2 XP_006710372.1 11 - 12 1219 145
XM_011540546.2 XP_011538848.1 13 - 14 1380 145
XM_017000137.1 XP_016855626.1 11 - 12 1585 145
XM_017000140.1 XP_016855629.1 10 - 11 1195 145
XM_017000144.1 XP_016855633.1 10 - 11 1103 145
GenBank mRNA AJ302735.1 - 11 - 12 1183 145
AJ347709.1 - 11 - 12 1184 145
BC001257.1 - 11 - 12 1184 145
U73704.1 - 11 - 12 1195 145

View Full Product Details