The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053274.2 | NP_444504.1 | 12 - 13 | 1254 | 245 |
NR_135089.1 | 12 - 13 | 1254 | 160 | ||
XM_005270401.3 | XP_005270458.1 | 11 - 12 | 1116 | 245 | |
XM_006710309.2 | XP_006710372.1 | 12 - 13 | 1256 | 245 | |
XM_011540546.2 | XP_011538848.1 | 14 - 15 | 1417 | 245 | |
XM_017000137.1 | XP_016855626.1 | 12 - 13 | 1622 | 245 | |
XM_017000140.1 | XP_016855629.1 | 11 - 12 | 1232 | 245 | |
XM_017000144.1 | XP_016855633.1 | 11 - 12 | 1140 | 245 | |
GenBank mRNA | AJ302735.1 | - | 12 - 13 | 1220 | 245 |
AJ347709.1 | - | 12 - 13 | 1221 | 245 | |
BC001257.1 | - | 12 - 13 | 1221 | 245 | |
U73704.1 | - | 12 - 13 | 1232 | 160 |