Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLMN
Entrez Gene ID
11146
Gene Name
glomulin, FKBP associated protein
Gene Aliases
FAP, FAP48, FAP68, FKBPAP, GLML, GVM, VMGLOM
Chromosome Location
Chr.1: 92245786 - 92441936 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319683.1 NP_001306612.1 16 - 17 1655 122
NM_053274.2 NP_444504.1 17 - 18 1697 122
NR_135089.1 16 - 17 1612 122
XM_005270401.3 XP_005270458.1 16 - 17 1559 122
XM_006710309.2 XP_006710372.1 17 - 18 1699 122
XM_011540546.2 XP_011538848.1 19 - 20 1860 122
XM_017000137.1 XP_016855626.1 17 - 18 2065 122
XM_017000138.1 XP_016855627.1 16 - 17 2023 122
XM_017000139.1 XP_016855628.1 16 - 17 1960 122
XM_017000140.1 XP_016855629.1 16 - 17 1675 122
XM_017000141.1 XP_016855630.1 16 - 17 1592 122
XM_017000142.1 XP_016855631.1 16 - 17 1657 122
XM_017000143.1 XP_016855632.1 16 - 17 2038 122
XM_017000144.1 XP_016855633.1 16 - 17 1583 122
GenBank mRNA AJ302735.1 - 17 - 18 1663 122
AJ347709.1 - 17 - 18 1664 122
AK295971.1 - 16 - 17 1633 122
BC001257.1 - 17 - 18 1664 122
BG187128.1 - 1 - 2 114 122

View Full Product Details