The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319683.1 | NP_001306612.1 | 1 - 2 | 85 | 75 |
NM_053274.2 | NP_444504.1 | 1 - 2 | 85 | 75 | |
NR_135089.1 | 1 - 2 | 85 | 75 | ||
XM_005270401.3 | XP_005270458.1 | 1 - 2 | 73 | 75 | |
XM_006710309.2 | XP_006710372.1 | 1 - 2 | 63 | 75 | |
XM_017000141.1 | XP_016855630.1 | 1 - 2 | 85 | 75 | |
XM_017000142.1 | XP_016855631.1 | 1 - 2 | 63 | 75 | |
XM_017000144.1 | XP_016855633.1 | 1 - 2 | 50 | 75 | |
GenBank mRNA | AJ302735.1 | - | 1 - 2 | 51 | 75 |
AJ347709.1 | - | 1 - 2 | 52 | 75 | |
AK295971.1 | - | 1 - 2 | 63 | 75 | |
BC001257.1 | - | 1 - 2 | 52 | 75 | |
DA729622.1 | - | 1 - 2 | 85 | 75 | |
U73704.1 | - | 1 - 2 | 63 | 75 |