The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243.4 | NP_001234.3 | 10 - 11 | 1379 | 94 |
NM_001281430.2 | NP_001268359.2 | 9 - 10 | 1291 | 94 | |
XM_011542441.2 | XP_011540743.1 | 10 - 11 | 1438 | 94 | |
XM_011542443.2 | XP_011540745.1 | 9 - 10 | 1456 | 94 | |
GenBank mRNA | BC136400.1 | - | 9 - 10 | 1198 | 94 |
BC171762.1 | - | 9 - 10 | 1198 | 94 | |
M83554.1 | - | 10 - 11 | 1379 | 94 | |
S75768.1 | - | 10 - 11 | 1157 | 94 |