The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136049.2 | NP_001129521.2 | 1 - 2 | 242 | 105 |
NM_033029.3 | NP_149018.2 | 1 - 2 | 242 | 105 | |
XM_011513270.1 | XP_011511572.1 | 1 - 2 | 225 | 105 | |
XM_011513271.1 | XP_011511573.1 | 1 - 2 | 225 | 105 | |
XM_017007428.1 | XP_016862917.1 | 1 - 2 | 225 | 105 | |
GenBank mRNA | AJ312399.1 | - | 1 - 2 | 242 | 105 |
AK299119.1 | - | 1 - 2 | 209 | 105 | |
AL832783.1 | - | 1 - 2 | 227 | 105 | |
AM920777.1 | - | 1 - 2 | 220 | 105 |