The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004446.2 | NP_004437.2 | 3 - 4 | 502 | 142 |
GenBank mRNA | AK296755.1 | - | 3 - 4 | 374 | 142 |
AK307534.1 | - | 1 - 2 | 76 | 142 | |
BC015494.1 | - | 3 - 4 | 290 | 142 | |
BC034797.1 | - | 3 - 4 | 298 | 142 | |
BC046156.1 | - | 3 - 4 | 502 | 142 | |
BC058921.1 | - | 3 - 4 | 339 | 142 | |
BC067841.1 | - | 3 - 4 | 374 | 142 | |
BC105983.1 | - | 3 - 4 | 300 | 142 | |
BC126275.1 | - | 3 - 4 | 370 | 142 | |
BC136465.1 | - | 3 - 4 | 270 | 142 | |
CR933648.1 | - | 3 - 4 | 341 | 142 | |
JQ080075.1 | - | 3 - 4 | 232 | 142 | |
X54326.1 | - | 1 - 2 | 74 | 142 |