The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004446.2 | NP_004437.2 | 6 - 7 | 888 | 74 |
GenBank mRNA | AK296755.1 | - | 6 - 7 | 760 | 74 |
AK307534.1 | - | 4 - 5 | 462 | 74 | |
AY493416.1 | - | 4 - 5 | 435 | 74 | |
BC015494.1 | - | 6 - 7 | 676 | 74 | |
BC034797.1 | - | 6 - 7 | 684 | 74 | |
BC046156.1 | - | 6 - 7 | 888 | 74 | |
BC058921.1 | - | 6 - 7 | 725 | 74 | |
BC105983.1 | - | 6 - 7 | 686 | 74 | |
BC126275.1 | - | 6 - 7 | 756 | 74 | |
BC136465.1 | - | 6 - 7 | 656 | 74 | |
CR933648.1 | - | 6 - 7 | 727 | 74 | |
JQ080075.1 | - | 6 - 7 | 618 | 74 | |
X07466.1 | - | 2 - 3 | 118 | 74 | |
X54326.1 | - | 4 - 5 | 460 | 74 |