Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR2A12
Entrez Gene ID
346525
Gene Name
olfactory receptor family 2 subfamily A member 12
Gene Aliases
OR2A12P, OR2A16P
Chromosome Location
Chr.7: 144095108 - 144096040 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004135.1 NP_001004135.1 1 - 1 54 108
GenBank mRNA BC136902.1 - 1 - 1 114 108
BC136903.1 - 1 - 1 114 108

View Full Product Details