Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TBKBP1
Entrez Gene ID
9755
Gene Name
TBK1 binding protein 1
Gene Aliases
ProSAPiP2, SINTBAD
Chromosome Location
Chr.17: 47694065 - 47712063 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014726.2 NP_055541.1 5 - 6 1663 159
XM_005257859.4 XP_005257916.1 6 - 7 962 159
XM_005257860.4 XP_005257917.1 6 - 7 1787 159
XM_005257861.4 XP_005257918.1 6 - 7 1267 159
XM_005257862.3 XP_005257919.1 6 - 7 887 159
XM_017025431.1 XP_016880920.1 5 - 6 2168 159
XM_017025432.1 XP_016880921.1 5 - 6 2167 159
GenBank mRNA AB018318.1 - 5 - 6 1663 159
BC152407.1 - 5 - 6 1663 159
BC167150.1 - 5 - 6 1663 159

View Full Product Details