The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014726.2 | NP_055541.1 | 6 - 7 | 1722 | 68 |
XM_005257859.4 | XP_005257916.1 | 7 - 8 | 1021 | 68 | |
XM_005257860.4 | XP_005257917.1 | 7 - 8 | 1846 | 68 | |
XM_005257861.4 | XP_005257918.1 | 7 - 8 | 1326 | 68 | |
XM_005257862.3 | XP_005257919.1 | 7 - 8 | 946 | 68 | |
XM_017025431.1 | XP_016880920.1 | 6 - 7 | 2227 | 68 | |
XM_017025432.1 | XP_016880921.1 | 6 - 7 | 2226 | 68 | |
GenBank mRNA | AB018318.1 | - | 6 - 7 | 1722 | 68 |
BC152407.1 | - | 6 - 7 | 1722 | 68 | |
BC167150.1 | - | 6 - 7 | 1722 | 68 |