Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM65
Entrez Gene ID
201292
Gene Name
tripartite motif containing 65
Gene Aliases
4732463G12Rik
Chromosome Location
Chr.17: 75880379 - 75897008 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256124.1 NP_001243053.1 2 - 3 578 55
NM_173547.3 NP_775818.2 2 - 3 578 55
XM_017024344.1 XP_016879833.1 2 - 3 580 55
GenBank mRNA AI242308.1 - 2 - 3 195 55
BC021259.2 - 2 - 3 546 55
BG753016.1 - 2 - 3 134 55
BQ070146.1 - - 409 55
DR002345.1 - 2 - 3 564 55

View Full Product Details