The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004669.2 | NP_004660.2 | 4 - 5 | 411 | 102 |
XM_017015281.1 | XP_016870770.1 | 4 - 5 | 616 | 102 | |
XM_017015282.1 | XP_016870771.1 | 4 - 5 | 579 | 102 | |
GenBank mRNA | AF102166.1 | - | 4 - 5 | 371 | 102 |
AI741256.1 | - | 2 - 3 | 102 | ||
BC007012.1 | - | 4 - 5 | 407 | 102 | |
BF439361.1 | - | 2 - 3 | 102 | ||
CR542066.1 | - | 3 - 4 | 295 | 102 |