The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001082533.1 | NP_001076002.1 | 8 - 9 | 1897 | 81 |
NM_001082534.1 | NP_001076003.1 | 8 - 9 | 1462 | 81 | |
NM_020178.4 | NP_064563.1 | 7 - 8 | 1771 | 81 | |
XM_017024878.1 | XP_016880367.1 | 5 - 6 | 625 | 81 | |
GenBank mRNA | AB001597.1 | - | 7 - 8 | 785 | 81 |
AB036836.1 | - | 8 - 9 | 1687 | 81 | |
AF288385.1 | - | 8 - 9 | 1453 | 81 | |
AK294652.1 | - | 6 - 7 | 1142 | 81 | |
AK313001.1 | - | 7 - 8 | 1347 | 81 | |
AL161812.1 | - | 7 - 8 | 787 | 81 | |
AY358509.1 | - | 7 - 8 | 1065 | 81 | |
BC020577.1 | - | 8 - 9 | 1429 | 81 | |
BC029865.1 | - | 8 - 9 | 1431 | 81 | |
BC047456.1 | - | 7 - 8 | 1684 | 81 | |
BC068462.1 | - | 7 - 8 | 1730 | 81 | |
CR749607.1 | - | 8 - 9 | 1233 | 81 |