Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
REXO4
Entrez Gene ID
57109
Gene Name
REX4 homolog, 3'-5' exonuclease
Gene Aliases
REX4, XPMC2, XPMC2H
Chromosome Location
Chr.9: 133406058 - 133418172 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001279350.1 NP_001266279.1 3 - 4 792 68
NM_001279351.1 NP_001266280.1 3 - 4 653 68
NM_020385.3 NP_065118.2 3 - 4 970 68
NR_103996.1 2 - 3 623 68
GenBank mRNA AF273304.1 - 3 - 4 918 68
AF295774.1 - 3 - 4 918 68
AK025493.1 - 3 - 4 824 68
AK295982.1 - 3 - 4 653 68
AK304547.1 - 3 - 4 708 68
AK314333.1 - 3 - 4 884 68
AK315904.1 - 4 - 5 643 68
AL136894.1 - 3 - 4 901 68
BC009274.2 - 3 - 4 873 68
BC071691.1 - 2 - 3 482 68

View Full Product Details