The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032279.3 | NP_115655.2 | 3 - 4 | 708 | 139 |
XM_011513232.2 | XP_011511534.1 | 3 - 4 | 455 | 139 | |
XM_017007318.1 | XP_016862807.1 | 3 - 4 | 344 | 139 | |
XM_017007319.1 | XP_016862808.1 | 3 - 4 | 455 | 139 | |
GenBank mRNA | AK095277.1 | - | 3 - 4 | 493 | 139 |
AY823162.1 | - | 3 - 4 | 385 | 139 | |
BC101496.1 | - | 3 - 4 | 479 | 139 |