The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002160.3 | NP_002151.2 | 13 - 14 | 4446 | 71 |
XM_005251972.3 | XP_005252029.1 | 13 - 14 | 4396 | 71 | |
XM_006717096.3 | XP_006717159.1 | 13 - 14 | 4396 | 71 | |
XM_011518625.2 | XP_011516927.1 | 13 - 14 | 4396 | 71 | |
XM_017014678.1 | XP_016870167.1 | 13 - 14 | 4396 | 71 | |
XM_017014680.1 | XP_016870169.1 | 13 - 14 | 4396 | 71 | |
GenBank mRNA | AB210038.1 | - | 13 - 14 | 4322 | 71 |
BC151843.1 | - | 13 - 14 | 4314 | 71 | |
M24630.1 | - | 11 - 12 | 2744 | 71 | |
M55618.1 | - | 12 - 13 | 4169 | 71 | |
X56160.1 | - | 12 - 13 | 4085 | 71 | |
X78565.1 | - | 13 - 14 | 4347 | 71 |