Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNC
Entrez Gene ID
3371
Gene Name
tenascin C
Gene Aliases
150-225, DFNA56, GMEM, GP, HXB, JI, TN, TN-C
Chromosome Location
Chr.9: 115019575 - 115118257 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002160.3 NP_002151.2 2 - 3 870 91
XM_005251972.3 XP_005252029.1 2 - 3 820 91
XM_005251973.3 XP_005252030.1 2 - 3 820 91
XM_005251974.3 XP_005252031.1 2 - 3 820 91
XM_005251975.3 XP_005252032.1 2 - 3 820 91
XM_006717096.3 XP_006717159.1 2 - 3 820 91
XM_006717097.3 XP_006717160.1 2 - 3 820 91
XM_006717098.3 XP_006717161.1 2 - 3 820 91
XM_006717101.3 XP_006717164.1 2 - 3 820 91
XM_011518625.2 XP_011516927.1 2 - 3 820 91
XM_011518626.2 XP_011516928.1 2 - 3 820 91
XM_011518628.2 XP_011516930.1 2 - 3 820 91
XM_011518629.2 XP_011516931.1 2 - 3 820 91
XM_017014678.1 XP_016870167.1 2 - 3 820 91
XM_017014679.1 XP_016870168.1 2 - 3 820 91
XM_017014680.1 XP_016870169.1 2 - 3 820 91
XM_017014681.1 XP_016870170.1 2 - 3 820 91
GenBank mRNA AB210038.1 - 2 - 3 746 91
AK304015.1 - 2 - 3 766 91
BC151843.1 - 2 - 3 741 91
CX871573.1 - 1 - 2 123 91
GU473235.1 - 1 - 2 458 91
M55618.1 - 1 - 2 593 91
X56160.1 - 1 - 2 512 91
X78565.1 - 2 - 3 771 91

View Full Product Details