The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004911.4 | NP_004902.1 | 6 - 7 | 1263 | 74 |
XM_006716185.2 | XP_006716248.1 | 6 - 7 | 1263 | 74 | |
GenBank mRNA | AK290971.1 | - | 6 - 7 | 1048 | 74 |
AK313299.1 | - | 6 - 7 | 1235 | 74 | |
BC000425.2 | - | 6 - 7 | 1025 | 74 | |
BC001928.1 | - | 6 - 7 | 1025 | 74 | |
BC006344.2 | - | 6 - 7 | 1213 | 74 | |
BC011754.2 | - | 6 - 7 | 1214 | 74 |