Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC41A3
Entrez Gene ID
54946
Gene Name
solute carrier family 41 member 3
Gene Aliases
SLC41A1-L2
Chromosome Location
Chr.3: 126006357 - 126101550 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008485.1 NP_001008485.1 9 - 10 1330 61
NM_001008486.1 NP_001008486.1 8 - 9 1222 61
NM_001008487.1 NP_001008487.1 8 - 9 1249 61
NM_001164475.1 NP_001157947.1 8 - 9 951 61
NM_017836.3 NP_060306.3 9 - 10 1330 61
XM_005247559.1 XP_005247616.1 9 - 10 1208 61
XM_005247562.1 XP_005247619.1 9 - 10 1224 61
XM_005247563.2 XP_005247620.1 9 - 10 1253 61
XM_005247564.1 XP_005247621.1 9 - 10 1208 61
XM_005247565.2 XP_005247622.1 8 - 9 1312 61
XM_006713681.2 XP_006713744.1 9 - 10 1170 61
XM_011512945.1 XP_011511247.1 8 - 9 1357 61
XM_017006705.1 XP_016862194.1 9 - 10 1197 61
XM_017006706.1 XP_016862195.1 9 - 10 1224 61
XM_017006707.1 XP_016862196.1 9 - 10 1247 61
XM_017006708.1 XP_016862197.1 8 - 9 1055 61
XM_017006709.1 XP_016862198.1 8 - 9 1110 61
GenBank mRNA AK000480.1 - 8 - 9 1053 61
AK021925.1 - 9 - 10 1016 61
AK022780.1 - 8 - 9 1051 61
AK091671.1 - 10 - 11 1134 61
AK093380.1 - 8 - 9 1246 61
AK094051.1 - 4 - 5 1339 61
AK298070.1 - 8 - 9 960 61
AW938624.1 - 2 - 3 206 61
BC009039.2 - 9 - 10 1163 61
BC009444.1 - 9 - 10 1148 61
BC028241.1 - 8 - 9 1146 61

View Full Product Details