Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNAH17
Entrez Gene ID
8632
Gene Name
dynein axonemal heavy chain 17
Gene Aliases
DNAHL1, DNEL2
Chromosome Location
Chr.17: 78423697 - 78577430 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173628.3 NP_775899.3 29 - 30 4803 109
XM_011525416.2 XP_011523718.1 29 - 30 4839 109

View Full Product Details