The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002976.3 | NP_002967.2 | 23 - 24 | 3840 | 99 |
NR_045628.1 | 24 - 25 | 3934 | 99 | ||
XM_006712680.2 | XP_006712743.1 | 23 - 24 | 3909 | 99 | |
XM_006712681.3 | XP_006712744.1 | 24 - 25 | 4134 | 99 | |
XM_006712682.3 | XP_006712745.1 | 25 - 26 | 4259 | 99 | |
XM_011511615.2 | XP_011509917.1 | 25 - 26 | 4205 | 99 | |
XM_017004667.1 | XP_016860156.1 | 25 - 26 | 4178 | 99 | |
GenBank mRNA | BC141864.1 | - | 24 - 25 | 3936 | 99 |
M91556.1 | - | 23 - 24 | 3849 | 99 |