The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005493.2 | NP_005484.2 | 6 - 7 | 1177 | 81 |
XM_006714945.2 | XP_006715008.1 | 6 - 7 | 657 | 81 | |
XM_011514205.2 | XP_011512507.1 | 6 - 7 | 1195 | 81 | |
XM_017010149.1 | XP_016865638.1 | 6 - 7 | 565 | 81 | |
GenBank mRNA | AB008515.1 | - | 6 - 7 | 866 | 81 |
AB055311.1 | - | 6 - 7 | 1177 | 81 | |
AF306510.1 | - | 6 - 7 | 715 | 81 | |
AK313334.1 | - | 5 - 6 | 568 | 81 | |
BC019886.1 | - | 6 - 7 | 631 | 81 | |
BC052781.1 | - | 6 - 7 | 1338 | 81 | |
BC063849.1 | - | 4 - 5 | 433 | 81 | |
KU921435.1 | - | 1 - 2 | 191 | 81 |