Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXO1
Entrez Gene ID
9156
Gene Name
exonuclease 1
Gene Aliases
HEX1, hExoI
Chromosome Location
Chr.1: 241847967 - 241889939 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003686.4 NP_003677.4 13 - 14 2717 152
GenBank mRNA AF042282.1 - 13 - 14 2621 152
AF091740.1 - 13 - 14 2614 152

View Full Product Details