Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHACTR1
Entrez Gene ID
221692
Gene Name
phosphatase and actin regulator 1
Gene Aliases
RPEL, RPEL1, dJ257A7.2
Chromosome Location
Chr.6: 12716635 - 13290484 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242648.2 NP_001229577.1 5 - 6 753 84
NM_001322308.1 NP_001309237.1 4 - 5 708 84
NM_001322309.1 NP_001309238.1 4 - 5 599 84
NM_001322310.1 NP_001309239.1 3 - 4 467 84
NM_001322311.1 NP_001309240.1 2 - 3 360 84
NM_001322312.1 NP_001309241.1 2 - 3 723 84
NM_001322313.1 NP_001309242.1 2 - 3 723 84
NM_001322314.1 NP_001309243.1 2 - 3 655 84
NM_030948.3 NP_112210.1 5 - 6 943 84
XM_005248934.2 XP_005248991.1 5 - 6 785 84
XM_017010452.1 XP_016865941.1 2 - 3 449 84
XM_017010454.1 XP_016865943.1 5 - 6 785 84
XM_017010455.1 XP_016865944.1 5 - 6 753 84
XM_017010456.1 XP_016865945.1 4 - 5 750 84
XM_017010457.1 XP_016865946.1 4 - 5 729 84
XM_017010458.1 XP_016865947.1 2 - 3 449 84
XM_017010459.1 XP_016865948.1 5 - 6 785 84
XM_017010460.1 XP_016865949.1 5 - 6 753 84
XM_017010461.1 XP_016865950.1 4 - 5 724 84
XM_017010462.1 XP_016865951.1 4 - 5 750 84
XM_017010463.1 XP_016865952.1 2 - 3 643 84
XM_017010464.1 XP_016865953.1 2 - 3 449 84
XM_017010465.1 XP_016865954.1 2 - 3 423 84
XM_017010466.1 XP_016865955.1 2 - 3 436 84
XM_017010467.1 XP_016865956.1 2 - 3 630 84
XM_017010469.1 XP_016865958.1 2 - 3 432 84
GenBank mRNA AB051520.1 - 4 - 5 550 84
AK090769.1 - 4 - 5 610 84
AK290017.1 - 5 - 6 604 84
AL598323.1 - 2 - 3 568 84
BC101526.1 - 4 - 5 517 84
BC101528.1 - 4 - 5 517 84

View Full Product Details