Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHACTR1
Entrez Gene ID
221692
Gene Name
phosphatase and actin regulator 1
Gene Aliases
RPEL, RPEL1, dJ257A7.2
Chromosome Location
Chr.6: 12716635 - 13290484 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242648.2 NP_001229577.1 11 - 12 1775 65
NM_001322308.1 NP_001309237.1 10 - 11 1730 65
NM_001322309.1 NP_001309238.1 10 - 11 1621 65
NM_001322310.1 NP_001309239.1 10 - 11 1696 65
NM_001322311.1 NP_001309240.1 8 - 9 1382 65
NM_001322312.1 NP_001309241.1 8 - 9 1745 65
NM_001322313.1 NP_001309242.1 9 - 10 1952 65
NM_001322314.1 NP_001309243.1 9 - 10 1884 65
NM_030948.3 NP_112210.1 11 - 12 1965 65
XM_005248934.2 XP_005248991.1 12 - 13 2014 65
XM_017010452.1 XP_016865941.1 9 - 10 1678 65
XM_017010454.1 XP_016865943.1 12 - 13 2014 65
XM_017010455.1 XP_016865944.1 12 - 13 1982 65
XM_017010456.1 XP_016865945.1 11 - 12 1979 65
XM_017010457.1 XP_016865946.1 11 - 12 1958 65
XM_017010458.1 XP_016865947.1 8 - 9 1471 65
XM_017010459.1 XP_016865948.1 11 - 12 1807 65
XM_017010460.1 XP_016865949.1 11 - 12 1775 65
XM_017010461.1 XP_016865950.1 10 - 11 1746 65
XM_017010462.1 XP_016865951.1 10 - 11 1772 65
XM_017010463.1 XP_016865952.1 9 - 10 1872 65
XM_017010464.1 XP_016865953.1 8 - 9 1471 65
XM_017010465.1 XP_016865954.1 9 - 10 1652 65
XM_017010466.1 XP_016865955.1 9 - 10 1665 65
XM_017010467.1 XP_016865956.1 8 - 9 1652 65
XM_017010469.1 XP_016865958.1 8 - 9 1454 65
GenBank mRNA AK090769.1 - 10 - 11 1632 65
AK290017.1 - 11 - 12 1626 65
BC101526.1 - 10 - 11 1539 65
BC101528.1 - 10 - 11 1539 65
BF026507.1 - - 146 65
BQ434462.1 - 1 - 2 182 65

View Full Product Details