Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC17A2
Entrez Gene ID
10246
Gene Name
solute carrier family 17 member 2
Gene Aliases
NPT3
Chromosome Location
Chr.6: 25912754 - 25930727 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286123.1 NP_001273052.1 10 - 11 1386 80
NM_005835.3 NP_005826.1 10 - 11 1688 80
XM_005248784.2 XP_005248841.1 11 - 12 1836 80
XM_006714949.3 XP_006715012.1 11 - 12 1573 80
XM_006714950.2 XP_006715013.1 10 - 11 1463 80
XM_017010159.1 XP_016865648.1 10 - 11 1726 80
GenBank mRNA BC104822.1 - 9 - 10 1211 80
BC112033.1 - 9 - 10 1211 80
BC143323.1 - 10 - 11 1357 80
U90544.1 - 10 - 11 1573 80

View Full Product Details