Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC17A2
Entrez Gene ID
10246
Gene Name
solute carrier family 17 member 2
Gene Aliases
NPT3
Chromosome Location
Chr.6: 25912754 - 25930727 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286123.1 NP_001273052.1 1 - 2 111 84
NM_001286125.1 NP_001273054.1 1 - 2 111 84
NM_005835.3 NP_005826.1 2 - 3 561 84
XM_005248784.2 XP_005248841.1 2 - 3 561 84
XM_006714949.3 XP_006715012.1 2 - 3 298 84
XM_006714951.1 XP_006715014.1 2 - 3 561 84
XM_017010160.1 XP_016865649.1 2 - 3 556 84
GenBank mRNA BC104822.1 - 1 - 2 84 84
BC112033.1 - 1 - 2 84 84
BC143323.1 - 1 - 2 82 84
U90544.1 - 2 - 3 446 84

View Full Product Details