Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC17A2
Entrez Gene ID
10246
Gene Name
solute carrier family 17 member 2
Gene Aliases
NPT3
Chromosome Location
Chr.6: 25912754 - 25930727 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286123.1 NP_001273052.1 6 - 7 855 93
NM_001286125.1 NP_001273054.1 6 - 7 855 93
NM_005835.3 NP_005826.1 7 - 8 1305 93
XM_005248784.2 XP_005248841.1 7 - 8 1305 93
XM_006714949.3 XP_006715012.1 7 - 8 1042 93
XM_006714950.2 XP_006715013.1 6 - 7 932 93
XM_006714951.1 XP_006715014.1 7 - 8 1305 93
XM_017010159.1 XP_016865648.1 6 - 7 1195 93
XM_017010160.1 XP_016865649.1 7 - 8 1300 93
GenBank mRNA BC104822.1 - 6 - 7 828 93
BC112033.1 - 6 - 7 828 93
BC143323.1 - 6 - 7 826 93
U90544.1 - 7 - 8 1190 93

View Full Product Details