The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008485.1 | NP_001008485.1 | 2 - 3 | 498 | 59 |
NM_017836.3 | NP_060306.3 | 2 - 3 | 498 | 59 | |
XM_005247559.1 | XP_005247616.1 | 2 - 3 | 331 | 59 | |
XM_005247562.1 | XP_005247619.1 | 2 - 3 | 392 | 59 | |
XM_005247563.2 | XP_005247620.1 | 2 - 3 | 421 | 59 | |
XM_005247564.1 | XP_005247621.1 | 2 - 3 | 331 | 59 | |
XM_006713681.2 | XP_006713744.1 | 2 - 3 | 338 | 59 | |
XM_017006705.1 | XP_016862194.1 | 2 - 3 | 320 | 59 | |
XM_017006706.1 | XP_016862195.1 | 2 - 3 | 392 | 59 | |
XM_017006707.1 | XP_016862196.1 | 2 - 3 | 415 | 59 | |
GenBank mRNA | AK021925.1 | - | 2 - 3 | 331 | 59 |
AK091671.1 | - | 3 - 4 | 302 | 59 | |
AK307107.1 | - | 2 - 3 | 376 | 59 | |
BC009039.2 | - | 2 - 3 | 331 | 59 | |
BC009444.1 | - | 2 - 3 | 316 | 59 | |
BC028241.1 | - | 1 - 2 | 314 | 59 |