The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282688.1 | NP_001269617.1 | 13 - 14 | 1986 | 87 |
NM_001282689.1 | NP_001269618.1 | 13 - 14 | 1868 | 87 | |
NM_001282690.1 | NP_001269619.1 | 13 - 14 | 2066 | 87 | |
NM_024077.4 | NP_076982.3 | 13 - 14 | 1989 | 87 | |
XM_005252196.2 | XP_005252253.1 | 13 - 14 | 1839 | 87 | |
XM_005252202.3 | XP_005252259.1 | 9 - 10 | 1221 | 87 | |
XM_006717282.2 | XP_006717345.1 | 13 - 14 | 1922 | 87 | |
XM_011519000.2 | XP_011517302.1 | 13 - 14 | 2043 | 87 | |
XM_011519001.1 | XP_011517303.1 | 12 - 13 | 1953 | 87 | |
XM_011519002.1 | XP_011517304.1 | 13 - 14 | 1929 | 87 | |
XM_011519003.1 | XP_011517305.1 | 13 - 14 | 1926 | 87 | |
XM_017015122.1 | XP_016870611.1 | 12 - 13 | 1866 | 87 | |
XM_017015123.1 | XP_016870612.1 | 13 - 14 | 1842 | 87 | |
XM_017015124.1 | XP_016870613.1 | 12 - 13 | 1749 | 87 | |
XM_017015125.1 | XP_016870614.1 | 11 - 12 | 1544 | 87 | |
GenBank mRNA | AB208940.1 | - | 13 - 14 | 1909 | 87 |
AF380995.1 | - | 13 - 14 | 1942 | 87 | |
AK023078.1 | - | 6 - 7 | 720 | 87 | |
AK090608.1 | - | 13 - 14 | 1956 | 87 | |
AK290182.1 | - | 13 - 14 | 1835 | 87 | |
AK299161.1 | - | 13 - 14 | 2077 | 87 | |
AK302852.1 | - | 13 - 14 | 1984 | 87 | |
AL136881.1 | - | 13 - 14 | 1822 | 87 | |
BC023142.1 | - | 7 - 8 | 836 | 87 | |
BC036109.1 | - | 13 - 14 | 1951 | 87 | |
BC068466.1 | - | 13 - 14 | 1924 | 87 |