The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282688.1 | NP_001269617.1 | 4 - 5 | 675 | 139 |
NM_001282689.1 | NP_001269618.1 | 4 - 5 | 557 | 139 | |
NM_001282690.1 | NP_001269619.1 | 4 - 5 | 755 | 139 | |
NM_024077.4 | NP_076982.3 | 4 - 5 | 678 | 139 | |
XM_005252196.2 | XP_005252253.1 | 4 - 5 | 645 | 139 | |
XM_006717282.2 | XP_006717345.1 | 4 - 5 | 611 | 139 | |
XM_011519000.2 | XP_011517302.1 | 4 - 5 | 732 | 139 | |
XM_011519001.1 | XP_011517303.1 | 4 - 5 | 732 | 139 | |
XM_011519002.1 | XP_011517304.1 | 4 - 5 | 732 | 139 | |
XM_011519003.1 | XP_011517305.1 | 4 - 5 | 732 | 139 | |
XM_017015122.1 | XP_016870611.1 | 4 - 5 | 645 | 139 | |
XM_017015123.1 | XP_016870612.1 | 4 - 5 | 645 | 139 | |
XM_017015124.1 | XP_016870613.1 | 4 - 5 | 645 | 139 | |
XM_017015125.1 | XP_016870614.1 | 2 - 3 | 271 | 139 | |
GenBank mRNA | AB208940.1 | - | 4 - 5 | 636 | 139 |
AF380995.1 | - | 4 - 5 | 631 | 139 | |
AK090608.1 | - | 4 - 5 | 645 | 139 | |
AK290182.1 | - | 4 - 5 | 524 | 139 | |
AK299161.1 | - | 4 - 5 | 766 | 139 | |
AK302852.1 | - | 4 - 5 | 673 | 139 | |
AL136881.1 | - | 4 - 5 | 511 | 139 | |
BC001189.1 | - | 4 - 5 | 511 | 139 | |
BC036109.1 | - | 4 - 5 | 640 | 139 | |
BC068466.1 | - | 4 - 5 | 613 | 139 | |
BC099900.1 | - | 4 - 5 | 614 | 139 | |
BX648787.1 | - | 2 - 3 | 3618 | 139 |