The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282688.1 | NP_001269617.1 | 8 - 9 | 1312 | 68 |
NM_001282689.1 | NP_001269618.1 | 8 - 9 | 1194 | 68 | |
NM_001282690.1 | NP_001269619.1 | 8 - 9 | 1392 | 68 | |
NM_024077.4 | NP_076982.3 | 8 - 9 | 1315 | 68 | |
XM_005252196.2 | XP_005252253.1 | 8 - 9 | 1165 | 68 | |
XM_005252202.3 | XP_005252259.1 | 4 - 5 | 547 | 68 | |
XM_006717282.2 | XP_006717345.1 | 8 - 9 | 1248 | 68 | |
XM_011519000.2 | XP_011517302.1 | 8 - 9 | 1369 | 68 | |
XM_011519002.1 | XP_011517304.1 | 8 - 9 | 1255 | 68 | |
XM_011519003.1 | XP_011517305.1 | 8 - 9 | 1252 | 68 | |
XM_017015123.1 | XP_016870612.1 | 8 - 9 | 1168 | 68 | |
XM_017015125.1 | XP_016870614.1 | 6 - 7 | 870 | 68 | |
GenBank mRNA | AB208940.1 | - | 8 - 9 | 1235 | 68 |
AF380995.1 | - | 8 - 9 | 1268 | 68 | |
AK023078.1 | - | 1 - 2 | 46 | 68 | |
AK090608.1 | - | 8 - 9 | 1282 | 68 | |
AK290182.1 | - | 8 - 9 | 1161 | 68 | |
AK299161.1 | - | 8 - 9 | 1403 | 68 | |
AK302852.1 | - | 8 - 9 | 1310 | 68 | |
AL136881.1 | - | 8 - 9 | 1148 | 68 | |
BC023142.1 | - | 2 - 3 | 162 | 68 | |
BC036109.1 | - | 8 - 9 | 1277 | 68 | |
BC068466.1 | - | 8 - 9 | 1250 | 68 |