The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000135.2 | NP_000126.2 | 1 - 2 | 124 | 157 |
NM_001018112.1 | NP_001018122.1 | 1 - 2 | 124 | 157 | |
NM_001286167.1 | NP_001273096.1 | 1 - 2 | 124 | 157 | |
XM_005256294.4 | XP_005256351.1 | 1 - 2 | 125 | 157 | |
XM_011522945.2 | XP_011521247.1 | 1 - 2 | 125 | 157 | |
XM_011522948.2 | XP_011521250.1 | 1 - 2 | 125 | 157 | |
XM_017023044.1 | XP_016878533.1 | 1 - 2 | 125 | 157 | |
XM_017023045.1 | XP_016878534.1 | 1 - 2 | 125 | 157 | |
XM_017023046.1 | XP_016878535.1 | 1 - 2 | 125 | 157 | |
GenBank mRNA | AK299282.1 | - | 1 - 2 | 113 | 157 |
BC008979.1 | - | 1 - 2 | 119 | 157 | |
BC064540.1 | - | 1 - 2 | 124 | 157 | |
BC120978.1 | - | 1 - 2 | 124 | 157 | |
BC120979.1 | - | 1 - 2 | 123 | 157 | |
BC127633.1 | - | 1 - 2 | 123 | 157 | |
BC141971.1 | - | 1 - 2 | 124 | 157 | |
BC141972.1 | - | 1 - 2 | 124 | 157 | |
X99226.1 | - | 1 - 2 | 113 | 157 |