The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319224.1 | NP_001306153.1 | 3 - 4 | 637 | 114 |
NM_003686.4 | NP_003677.4 | 2 - 3 | 474 | 114 | |
NM_006027.4 | NP_006018.4 | 2 - 3 | 474 | 114 | |
NM_130398.3 | NP_569082.2 | 4 - 5 | 742 | 114 | |
XM_006711840.2 | XP_006711903.1 | 4 - 5 | 745 | 114 | |
XM_011544321.1 | XP_011542623.1 | 3 - 4 | 639 | 114 | |
XM_011544322.1 | XP_011542624.1 | 3 - 4 | 708 | 114 | |
XM_011544323.2 | XP_011542625.1 | 4 - 5 | 745 | 114 | |
GenBank mRNA | AF042282.1 | - | 2 - 3 | 378 | 114 |
AF060479.1 | - | 2 - 3 | 369 | 114 | |
AF084974.1 | - | 2 - 3 | 369 | 114 | |
AF091740.1 | - | 2 - 3 | 371 | 114 | |
AK291291.1 | - | 3 - 4 | 625 | 114 | |
BC007491.2 | - | 4 - 5 | 690 | 114 | |
CD644038.1 | - | 3 - 4 | 605 | 114 |