The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004759.4 | NP_004750.1 | 1 - 2 | 490 | 121 |
NM_032960.3 | NP_116584.2 | 1 - 2 | 490 | 121 | |
XM_005273353.3 | XP_005273410.1 | 1 - 2 | 598 | 121 | |
GenBank mRNA | BC036060.1 | - | 1 - 2 | 490 | 121 |
BC052584.1 | - | 1 - 2 | 473 | 121 |